Skip to Content
Merck
All Photos(1)

Key Documents

Z381535

Heating blocks for Techne® Dri-Block® heater

holds 1 × Falcon 96-well round bottom plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NB.76

feature

Holds 1 x Falcon 96-well round bottom plate

manufacturer/tradename

Techne F4468

Looking for similar products? Visit Product Comparison Guide

General description

Interchangeable blocks machined of aluminum for rapid heat transfer. Close tolerances assure efficient, rapid heating. Each block has a threaded thermometer hole for independent temperature monitoring, which also fits the block extraction tool.

Compatibility

For use with Model DB-1M only.

Legal Information

Dri-Block is a registered trademark of Techne, Inc.
Techne is a registered trademark of Techne, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie Mok et al.
Nature communications, 11(1), 4757-4757 (2020-09-23)
Local tissue mechanics play a critical role in cell function, but measuring these properties at cellular length scales in living 3D tissues can present considerable challenges. Here we present thermoresponsive, smart material microgels that can be dispersed or injected into tissues
Hamid Bolouri et al.
Scientific reports, 10(1), 1915-1915 (2020-02-07)
Failure to clear antigens causes CD8+ T cells to become increasingly hypo-functional, a state known as exhaustion. We combined manually extracted information from published literature with gene expression data from diverse model systems to infer a set of molecular regulatory
Irene Cassaniti et al.
Vector borne and zoonotic diseases (Larchmont, N.Y.), 20(4), 295-302 (2019-12-11)
Dengue virus (DENV), a member of the family Flaviviridae, is the causative agent of dengue fever, the most prevalent mosquito-borne viral illness in humans, representing a major public health concern in the tropical countries. Although humoral immunity to DENV has
Aurelien B L Colamartino et al.
Frontiers in immunology, 10, 2873-2873 (2020-01-11)
NK-cell resistance to transduction is a major technical hurdle for developing NK-cell immunotherapy. By using Baboon envelope pseudotyped lentiviral vectors (BaEV-LVs) encoding eGFP, we obtained a transduction rate of 23.0 ± 6.6% (mean ± SD) in freshly-isolated human NK-cells (FI-NK)
Sofia E M Andersson et al.
Immunity & ageing : I & A, 17, 12-12 (2020-05-30)
Exercise can improve immune health and is beneficial for physical function in patients with rheumatoid arthritis (RA), but the immunological mechanisms are largely unknown. We evaluated the effect of moderate- to high intensity exercise with person-centred guidance on cells of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service