Z375233
Masterflex™ pump drive
digital standard drive, AC/DC input 190 - 260 V AC
All Photos(1)
About This Item
UNSPSC Code:
41100000
Recommended Products
manufacturer/tradename
Cole Parmer
General description
Does not include pump head.
Economy: unidirectional, lowest priced variable-speed drive, ± 5% speed uniformity. Accepts one EASY-LOAD head
Precision: bidirectional, soft start, 10-turn speed control, ± 1% speed uniformity. Accepts DB9 cable for remote control capabilities
Digital: bidirectional, high-precision digital speed control, ± 0.25% speed uniformity. Displays rpm, flow rate, and % speed. Accepts DB9 cable for remote control capabilities
Economy: unidirectional, lowest priced variable-speed drive, ± 5% speed uniformity. Accepts one EASY-LOAD head
Precision: bidirectional, soft start, 10-turn speed control, ± 1% speed uniformity. Accepts DB9 cable for remote control capabilities
Digital: bidirectional, high-precision digital speed control, ± 0.25% speed uniformity. Displays rpm, flow rate, and % speed. Accepts DB9 cable for remote control capabilities
Legal Information
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)