Recommended Products
material
polypropylene
red
feature
autoclavable at 121 °C (2 bar), acc to DIN EN 285
positions 8 × 16
Holds 128 x microtubes
manufacturer/tradename
BRAND 4341052
parameter
−20-+90 °C temperature
L × W × H
265 mm × 126 mm × 38 mm
tube diam.
11 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Microcentrifuge tube racks are stackable, with alphanumerical positions. Racks are made of PP (filled, density 1.2 g/cm), will not float in waterbath. Sturdy construction racks are supplied in two-pieces for convenient and permanent assembly.
Other Notes
BR4341052 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service