Key Documents
WPB01392WA
Whirl-Pak® Speci-Sponge® Bag with Sterile Gloves
532 mL capacity(18 oz), pack of 100 ea
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
description
with Sterile Glove
packaging
pack of 100 ea
manufacturer/tradename
Nasco B01392WA
W × L × thickness
11.5 cm × 23 cm × 0.064 mm
capacity
532 mL × 18 oz
532 mL capacity(18 oz)
General description
Includes a sterile polyethylene glove attached in a bottom compartment. To use the glove, tear off the glove pack along the perforation. The glove is folded with the open end slightly extended so the glove can be held and shaken open without touching the hand area.
Designed for environmental surface sampling of work areas, equipment, animal carcasses, and any other place where testing for Listeria, Salmonella, E.coli, coliforms, and other foodborne pathogens, or anthrax is required.
Legal Information
Speci-Sponge is a registered trademark of The Aristotle Corporation
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)