Skip to Content
Merck
All Photos(1)

Key Documents

WH0196403M1

Sigma-Aldrich

Monoclonal Anti-DTX3 antibody produced in mouse

clone 4F10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FLJ34766, Anti-RNF154, Anti-deltex 3 homolog (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€490.00

€490.00


Check Cart for Availability


Select a Size

Change View
100 μG
€490.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€490.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG3λ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DTX3(196403)

General description

Deltex 3 homolog (DTX3) has a truncated domain 1 and a proline-rich region. The gene encoding it is localized on human chromosome 12q13.3.

Immunogen

DTX3 (NP_848597, 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MCGRFYGQLVGNQPQNGRMLVSKDATLLLPSYEKYGTIVIQYVFPPGVQGAEHPNPGVRYPGTTRVAYLPDCPEGNKVLTLFRKAFDQRLTFTIGTSMTT

Biochem/physiol Actions

Deltex 3 homolog (DTX3) acts as an E3 ubiquitin ligase.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kunihiko Takeyama et al.
The Journal of biological chemistry, 278(24), 21930-21937 (2003-04-03)
Members of the DTX (Deltex) family act as Notch signaling modifiers and may also regulate transcription through interactions with specific transcription factors. DTX proteins have a basic N terminus; a central proline-rich region; and a C-terminal RING finger domain, a
Kristian Almstrup et al.
Cell and tissue research, 322(1), 159-165 (2005-04-23)
Testicular cancer is the most common malignancy among men in the reproductive age and the incidence is increasing, probably caused by environmental factors. Most testicular cancers are testicular germ cell tumours and all originate from a carcinoma in situ (CIS)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service