Skip to Content
Merck
All Photos(1)

Key Documents

SRP6587

Sigma-Aldrich

Histone H4 (1-103) human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

11.5 kDa (1-136 aa)

packaging

pkg of 100 and 250 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HIST1H4A(8359)

General description

H2A is a core component of nucleosome. A nucleosome is the basic repeating unit of chromatin in which 146 base pairs of DNA wrap twice around an octamer of histones. The octamer is composed of two of each histone H2A, H2B, H3, and H4. DNA accessibility is regulated via a complex set of post-translational modifications of these histones, also called histone code, and nucleosome remodeling. Histones H2A and H2B form a dimer. Histones H3 and H4 form a tetramer. The combination of two H2A/H2B dimers and one H3/H4 tetramer create the nucleosome core. Histone H4 undergoes many modifications which include acetylation, phosphorylation and methylation that are important for regulation of gene transcription. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability.

Physical form

Lyophilized powder. Recommended buffer is 50 mM NaPO₄ containing 100 mM sodium chloride and 20% glycerol.

Reconstitution

Resuspend in buffer of choice.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service