Skip to Content
Merck
All Photos(1)

Key Documents

SRP6498

Sigma-Aldrich

PDGFRb/CD140b human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CD-140B, JTK-12, JTK12, PDGF-R-β, PDGF-R-beta, PDGFR, PDGFR-β, PDGFR-1, PDGFR-B, PDGFR-beta, PDGFR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 57.1 kDa
observed mol wt 85-95 kDa by SDS-PAGE (DTT-reduced.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PDGFRB(5159)

General description

Human platelet-derived growth factor receptor, β polypeptide (PDGFRB), also called J03278, M21616, CD140B, JTK12, PDGF-R-β and PDGFR, is receptor that binds specifically to PDGFB and has a tyrosine-protein kinase activity. Is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor (PDGF) family. The PDGFR/PDGF system includes two receptors (PDGFRA and PDGFRB) and four ligands (A, B, C and D). The receptors PDGFRA and PDGFRB are related in sequence and both are members of the class III subtype of receptor tyrosine kinases (RTKs). Other class III RTKs are CSF1R, KIT and FLT3. PDGF binding induces receptor homo- and heterodimerization and signal transduction. The expression of the α and β receptors is independently regulated in various cell types. Recombinant soluble PDGFRB binds PDGF with high affinity and is potent PDGF antagonist. The ligands form either homo- or heterodimers (PDGF-AA, -AB, -BB, -CC, -DD). The four PDGFs are inactive in their monomeric forms. The PDGFs bind to the protein tyrosine kinase receptors PDGF receptor-α and -β. These two receptor isoforms dimerize upon binding the PDGF dimer, leading to three possible receptor combinations, namely -αα, -ββ and -αβ. PDGF-CC specifically interacts with PDGFR-αα and -αβ, but not with –ββ, and thereby resembles PDGF-AB. PDGF-DD binds to PDGFR-ββ with high affinity and to PDGFR-αβ to a much lower extent and is regarded as PDGFR-ββ specific. PDGF-AA binds only to PDGFR-αα, while PDGF-BB is the only PDGF that can bind all three receptor combinations with high affinity.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service