Skip to Content
Merck
All Photos(1)

Key Documents

SRP6447

Sigma-Aldrich

CNTN2/Contactin-2 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

AXT, Contactin-2, DKFZp781D102, FLJ37193, FLJ42746, MGC157722, TAG-1, TAX, TAX-1, TAX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 109 kDa
observed mol wt 140 kDa by SDS-PAGE (reducing) (Ser 31 is the predicted N-terminal .)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CNTN2(6900)

General description

Contactin-2 also known as CNTN2, TAX1 (transiently-expressed axonal glycoprotein), TAG1 (transient axonal glycoprotein), and axonin-1, and is a member of the immunoglobulin superfamily. CNTN2 consists of six Ig-like domains and four fibronectin type III domains, and is anchored to the membrane by glycosylphosphatidylinositol (GPI), whereas the soluble form can be released by a GPI-specific phospholipase. As a neural cell adhesion molecule expressed by a subset of neuronal populations in the developing CNS and PNS, CNTN2 mediates cell-cell interactions either via homophilic, or heterophilic contacts with various adhesion molecules including NgCAM, NrCAM, NCAM and neurocan. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. It may also be involved in glial tumorigenesis and may provide a potential target for therapeutic intervention.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service