Skip to Content
Merck
All Photos(1)

Documents

SRP0649

Sigma-Aldrich

ARG2, His-Tag

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

Kidney-Type Arginase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in baculovirus infected Sf9 cells

form

frozen liquid

mol wt

38 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ARG2(384)

General description

Human Arginase 2B (ARG2B) protein, also known as LCN7 and LIECF3, GenBank Accession No. NM_001172, amino acids 22-355, with an N-terminal His-tag, expressed in an SF9 insect cell expression system. MW = 38 kDa.

Physical form

Aqueous solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service