Skip to Content
Merck
All Photos(1)

Key Documents

SRP0379

Sigma-Aldrich

EZH2 human

recombinant, expressed in baculovirus infected Sf9 cells, ≥49% (SDS-PAGE)

Synonym(s):

ENX1, KMT6, enhancer of zeste homolog 2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€648.00

€648.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
20 μG
€648.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€648.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥49% (SDS-PAGE)

form

aqueous solution

mol wt

114 kDa

packaging

pkg of 20 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EZH2(2146)

General description

Human EZH2 (GenBank Accession No. NM_004456), amino acids 2-end with N-terminal His-GST-tag, MW= 114 kDa, expressed in a Sf9 Baculovirus expression system.

Physical form

Formulated in 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 16 mM glutathione, 20% glycerol and 3 mM DTT.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Zhang et al.
Nucleic acids research, 45(16), 9348-9360 (2017-06-24)
In eukaryotic cells, the gene expression status is strictly controlled by epigenetic modifications on chromatin. The repressive status of chromatin largely contributes to HIV latency. Studies have shown that modification of histone H3K27 acts as a key molecular switch for
Rakotobe, D. et al.
Journal of Virology, 5, 32-32 (2008)
Sooryanarayana Varambally et al.
Science (New York, N.Y.), 322(5908), 1695-1699 (2008-11-15)
Enhancer of zeste homolog 2 (EZH2) is a mammalian histone methyltransferase that contributes to the epigenetic silencing of target genes and regulates the survival and metastasis of cancer cells. EZH2 is overexpressed in aggressive solid tumors by mechanisms that remain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service