Skip to Content
Merck
All Photos(1)

Documents

SML3149

Sigma-Aldrich

BI8626

≥98% (HPLC)

Synonym(s):

BI 8626, BI-8626, N-(3-(Aminomethyl)benzyl)-8-(4-benzylpiperazin-1-yl)pyrimido[5,4-d]pyrimidin-4-amine, N1-[8-[4-(Phenylmethyl)-1-piperazinyl]pyrimido[5,4-d]pyrimidin-4-yl]-1,3-benzenedimethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28N8
CAS Number:
Molecular Weight:
440.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

BI8626 is a selective HUWE1 (HECTH9; MULE; ARF-BP1) inhibitor (IC50 = 0.9 µM by HUWE1 HECT-domain auto-ubiquitination assay; IC50 >50 µM against other HECT-domain Ub ligases tested). BI8626 inhibits the degradation of HUWE1 target proteiins, including MCL1, MIZ1, and TopBP1 (10-20 µM). BI8626 suppresses Ls174T colony formation (IC50 = 0.7 µM in 5 days) and the growth of three colon carcinoma cell lines (20 µM for 5 days; HCT116, HT29, SW480).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E K Es'kov
Biofizika, 20(4), 646-651 (1975-07-01)
Bee phonoreceptors are revealed by electrophysiological method. These are hair sensillae located between facet eyes and occipital commissure. The region of the highest sensitivity of receptors in within the range of the maximum energy of various sound signals used in
Stefanie Peter et al.
EMBO molecular medicine, 6(12), 1525-1541 (2014-09-26)
Deregulated expression of MYC is a driver of colorectal carcinogenesis, necessitating novel strategies to inhibit MYC function. The ubiquitin ligase HUWE1 (HECTH9, ARF-BP1, MULE) associates with both MYC and the MYC-associated protein MIZ1. We show here that HUWE1 is required
Aswini Krishnan et al.
Nature communications, 11(1), 2056-2056 (2020-04-30)
Papillary thyroid cancer (PTC) is the most common type of endocrine malignancy. By RNA-seq analysis, we identify a RET rearrangement in the tumour material of a patient who does not harbour any known RAS or BRAF mutations. This new gene
Theresa Endres et al.
Molecular cell, 81(4), 830-844 (2021-01-17)
The MYC oncoprotein globally affects the function of RNA polymerase II (RNAPII). The ability of MYC to promote transcription elongation depends on its ubiquitylation. Here, we show that MYC and PAF1c (polymerase II-associated factor 1 complex) interact directly and mutually

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service