Skip to Content
Merck
All Photos(1)

Key Documents

SML2575

Sigma-Aldrich

GSHtracer

≥98% (HPLC)

Synonym(s):

2-Cyano-N,N-dimethyl-3-(2,3,6,7-tetrahydro-11-oxo-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-10-yl)-2-propenamide, Fluorescent real-time thiol tracer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3O3
CAS Number:
Molecular Weight:
363.41
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

faint red to very dark red

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1OC2=C3C4=C(CCCN4CCC3)C=C2C=C1/C=C(C#N)/C(N(C)C)=O

Biochem/physiol Actions

GSHtracer is a real-time probe monitoring of GSH (reduced glutathione) dynamics and heterogeneity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eui Man Jeong et al.
Stem cell reports, 10(2), 600-614 (2018-01-09)
The core functions of stem cells (SCs) are critically regulated by their cellular redox status. Glutathione is the most abundant non-protein thiol functioning as an antioxidant and a redox regulator. However, an investigation into the relationship between glutathione-mediated redox capacity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service