Skip to Content
Merck
All Photos(1)

Documents

SML1953

Sigma-Aldrich

GDC-0879

≥98% (HPLC)

Synonym(s):

1H-Inden-1-one, 2,3-dihydro-5-[1-(2-hydroxyethyl)-3-(4-pyridinyl)-1H-pyrazol-4-yl]-, oxime, 2,3-Dihydro-5-[1-(2-hydroxyethyl)-3-(4-pyridinyl)-1H-pyrazol-4-yl]-1H-inden-1-one oxime, 2-[4-[(1E)-1-(Hydroxyimino)-2,3-dihydro-1H-inden-5-yl]-3-(pyridine-4-yl)-1H-pyrazol-1-yl]ethan-1-ol, 5-(1-(2-Hydroxyethyl)-3-(pyridin-4-yl)-1H-pyrazol-4-yl)-2,3-dihydro-1H-inden-1-one oxime, AR00341677, GDC0879

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N4O2
CAS Number:
Molecular Weight:
334.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OCCN(N=C1C2=CC=NC=C2)C=C1C3=CC=C4C(CC/C4=N\O)=C3

InChI

1S/C19H18N4O2/c24-10-9-23-12-17(19(21-23)13-5-7-20-8-6-13)15-1-3-16-14(11-15)2-4-18(16)22-25/h1,3,5-8,11-12,24-25H,2,4,9-10H2/b22-18+

InChI key

DEZZLWQELQORIU-RELWKKBWSA-N

Biochem/physiol Actions

GDC-0879 (AR00341677) is an orally active, ATP site-targeting, highly potent Raf inhibitor (IC50 against V600E B-Raf-catalyzed MEK phophorylation = 0.13 nM with 4 μM ATP; IC50 against pERK in V600E B-Raf-expressing cells = 29 nM/Colo205, 59 nM/A375, 63 nM/Malme-3M) with great selectivity over >200 protein kinases (IC50 >1 μM). Oral administration is shown to effectively suppress Colo205 and A375 xenografts-derived tumor growth and pMEK1 in mice in vivo (35 mg/kg bid; 50-200 mg/kg qd or bid). Paradoxically, GDC-0879 also induces wild-type and enhances V600E B-Raf dimerization in an allosteric manner independent of kinase activity inhibition, resulting in selective anti-proliferation activity against V600E B-Raf cancer cells, but not in KRAS mutant or RAS/WT Raf cells, where GDC-0879 induces formation of B–C and A–B heterodimers and downstream signaling activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service