Skip to Content
Merck
All Photos(1)

Documents

SML1625

Sigma-Aldrich

Gemifloxacin mesylate

≥98% (HPLC)

Synonym(s):

7-(4-Aminomethyl-3-methyloxyiminopyrrolidin-1-yl)-1-cyclopropyl-6-fluoro-4-oxo-1, 4-dihydro-1,8-naphthyridine-3-carboxylic acid mesylate, 7-[(4Z)-3-(Aminomethyl)-4-(methoxyimino)-1-pyrrolidinyl]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid monomethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20FN5O4 · CH4O3S
CAS Number:
Molecular Weight:
485.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

H2O: 10 mg/mL, clear

storage temp.

room temp

SMILES string

CS(=O)(O)=O.OC(C1=CN(C2CC2)C3=NC(N4C/C(C(CN)C4)=N\OC)=C(F)C=C3C1=O)=O

InChI

1S/C18H20FN5O4.CH4O3S/c1-28-22-14-8-23(6-9(14)5-20)17-13(19)4-11-15(25)12(18(26)27)7-24(10-2-3-10)16(11)21-17;1-5(2,3)4/h4,7,9-10H,2-3,5-6,8,20H2,1H3,(H,26,27);1H3,(H,2,3,4)/b22-14+;

InChI key

JIYMVSQRGZEYAX-CWUUNJJBSA-N

Biochem/physiol Actions

Gemifloxacin is a broad-spectrum quinolone antibacterial. Fluoroquinolones stabilize DNA strand breaks created by DNA gyrase and topoisomerase IV by binding to the enzyme-DNA complex generating persistent, covalent enzyme–DNA adducts, inhibiting DNA synthesis. Gemifloxacin is used in the treatment of acute bacterial exacerbation of chronic bronchitis and mild-to-moderate pneumonia. Gemifloxacin has also been found to inhibit migration and invasion of human colon cancer cells.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2 - STOT RE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service