Skip to Content
Merck
All Photos(1)

Documents

SML1552

Sigma-Aldrich

Tipiracil hydrochloride

≥98% (HPLC)

Synonym(s):

5-Chloro-6-[(2-imino-1-pyrrolidinyl)methyl]-2,4(1H,3H)-pyrimidinedione hydrochloride, TPI hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11ClN4O2 · HCl
CAS Number:
Molecular Weight:
279.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[H]Cl.ClC1=C(CN2C(CCC2)=N)NC(NC1=O)=O

InChI

1S/C9H11ClN4O2.ClH/c10-7-5(12-9(16)13-8(7)15)4-14-3-1-2-6(14)11;/h11H,1-4H2,(H2,12,13,15,16);1H

InChI key

KGHYQYACJRXCAT-UHFFFAOYSA-N

Gene Information

human ... TYMP(1890)

Biochem/physiol Actions

Tipiracil is an inhibitor of thymidine phosphorylase. Tipiracil is used in combination with trifluridine as TAS-102 for the treatment of refractory metastatic colorectal cancer. Tipiracil increases the bioavailability of trifluridine by blocking the enzyme that would otherwise protect the tumors by metabolizing trifluridine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kei Higashikawa et al.
Nuclear medicine and biology, 82-83, 25-32 (2019-12-24)
Although liver biopsy is the gold standard for the diagnosis of nonalcoholic steatohepatitis (NASH), it has several problems including high invasiveness and sampling errors. Therefore, the development of alternative methods to overcome these disadvantages is strongly required. In this study
Sumaira Javaid et al.
PloS one, 14(11), e0225056-e0225056 (2019-11-20)
Natural flora is the richest source of novel therapeutic agents due to their immense chemical diversity and novel biological properties. In this regard, eighteen natural products belonging to different chemical classes were evaluated for their thymidine phosphorylase (TP) inhibitory activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service