Skip to Content
Merck
All Photos(1)

Key Documents

SML1361

Sigma-Aldrich

BRD73954

≥98% (HPLC)

Synonym(s):

N-Hydroxy-N′-phenethyl-isophthalamide, N1-Hydroxy-N3-(2-phenylethyl)-1,3-benzenedicarboxamide, N1-Hydroxy-N3-phenethylisophthalamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2O3
CAS Number:
Molecular Weight:
284.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

ONC(C1=CC(C(NCCC2=CC=CC=C2)=O)=CC=C1)=O

InChI

1S/C16H16N2O3/c19-15(17-10-9-12-5-2-1-3-6-12)13-7-4-8-14(11-13)16(20)18-21/h1-8,11,21H,9-10H2,(H,17,19)(H,18,20)

InChI key

FIHKWEQJEDRIFS-UHFFFAOYSA-N

Biochem/physiol Actions

BRD73954 is the first discovered histone deacetylase 6/8 dual inhibitor. BRD73954 has an IC50 of 36 nM for the class IIb HDAC6 and 120 nM for the class I HDAC8 with IC450′s ranging from 9 μM to greater than 33 μM for other HDACs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service