Skip to Content
Merck
All Photos(1)

Documents

SML1359

Sigma-Aldrich

CM037

≥97% (HPLC)

Synonym(s):

2-[[3,4-Dihydro-4-oxo-3-[3-(1-pyrrolidinyl)propyl][1]benzothieno[3,2-d]pyrimidin-2-yl]thio]-acetic acid ethyl ester, A37, Ethyl 2-((4-oxo-3-(3-(pyrrolidin-1-yl)propyl)-3,4-dihydrobenzo[4,5]thieno[3,2-d]pyrimidin-2-yl)thio)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N3O3S2
CAS Number:
Molecular Weight:
431.57
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(CCCN2CCCC2)C(SCC(OCC)=O)=NC3=C1SC4=CC=CC=C43

InChI

1S/C21H25N3O3S2/c1-2-27-17(25)14-28-21-22-18-15-8-3-4-9-16(15)29-19(18)20(26)24(21)13-7-12-23-10-5-6-11-23/h3-4,8-9H,2,5-7,10-14H2,1H3

InChI key

SKDRHRAYBYQVNU-UHFFFAOYSA-N

Biochem/physiol Actions

CM037 is a potent and selective inhibitor of human aldehyde dehydrogenase 1A1 (ALDH1A1). CM037 binds to the aldehyde binding pocket of ALDH1A1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service