Skip to Content
Merck
All Photos(1)

Key Documents

SML1077

Sigma-Aldrich

ML298

≥98% (HPLC)

Synonym(s):

3,4-Difluoro-N-[2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]dec-8-yl]ethyl]-benzamide, CID53393915

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€114.00
25 MG
€360.00

€114.00


Estimated to ship onMay 07, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€114.00
25 MG
€360.00

About This Item

Empirical Formula (Hill Notation):
C22H23F3N4O2
CAS Number:
Molecular Weight:
432.44
UNSPSC Code:
41106300
NACRES:
NA.77

€114.00


Estimated to ship onMay 07, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)C(=O)NCCN2CCC3(N(CNC3=O)c4cc(ccc4)F)CC2)F

InChI

1S/C22H23F3N4O2/c23-16-2-1-3-17(13-16)29-14-27-21(31)22(29)6-9-28(10-7-22)11-8-26-20(30)15-4-5-18(24)19(25)12-15/h1-5,12-13H,6-11,14H2,(H,26,30)(H,27,31)

InChI key

XAPVAKKLQGLNOY-UHFFFAOYSA-N

Biochem/physiol Actions

ML298 is a Phospholipase D2 (PLD2) inhibitor.
ML298 is a potent, specific inhibitor of Phospholipase D2 (PLD2; IC50 = 355 nM). ML298 does not affect PLD1 activity at concentrations up to 20 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service