Skip to Content
Merck
All Photos(1)

Key Documents

SML0587

Sigma-Aldrich

PF9601N

≥98% (HPLC)

Synonym(s):

5-(Phenylmethoxy)-N-2-propynyl- 1H-Indole-2-methanamine, FA-73, N-(2-Propynyl)-2-(5-benzyloxy-indolyl) methylamine, PF 9601N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O
CAS Number:
Molecular Weight:
290.36
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PF9601N is a selective and potent monoamine oxidase B inhibitor that exhibit anti-Parkinsonian effects in several models of PD.
PF9601N is a selective and potent monoamine oxidase B inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Our company offers many products related to histamine synthesis and metabolism for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service