Skip to Content
Merck
All Photos(1)

Key Documents

SML0413

Sigma-Aldrich

FQI1

≥98% (HPLC)

Synonym(s):

8-(2-Ethoxyphenyl)-7,8-dihydro-1,3-Dioxolo[4,5-g]quinolin-6(5H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€98.60
25 MG
€267.00

€98.60


Estimated to ship onMay 13, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€98.60
25 MG
€267.00

About This Item

Empirical Formula (Hill Notation):
C18H17NO4
CAS Number:
Molecular Weight:
311.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€98.60


Estimated to ship onMay 13, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

O=C1CC(C2=CC=CC=C2OCC)C3=CC4=C(OCO4)C=C3N1

InChI

1S/C18H17NO4/c1-2-21-15-6-4-3-5-11(15)12-8-18(20)19-14-9-17-16(7-13(12)14)22-10-23-17/h3-7,9,12H,2,8,10H2,1H3,(H,19,20)

InChI key

YKSYGLXHLSPWLB-UHFFFAOYSA-N

Biochem/physiol Actions

FQI1 inhibits the DNA-binding activity of LSF, a ubiquitous transcription factor that is up regulated in many hepatocellular carcinomas (HCC). FQI1 inhibits LSF-dependent luciferase reporter expression, blocks proliferation in cancer cell lines, and induces apoptosis in liver cancer cells but is not toxic to primary hepatocytes. FQI1 also inhibits tumor growth in a mouse HCC xenograft model.
FQI1 is an inhibitor of transcription factor LSF.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L S Willoughby et al.
BMC cancer, 20(1), 552-552 (2020-06-17)
The oncogene LSF (encoded by TFCP2) has been proposed as a novel therapeutic target for multiple cancers. LSF overexpression in patient tumors correlates with poor prognosis in particular for both hepatocellular carcinoma and colorectal cancer. The limited treatment outcomes for
Seung Han Son et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 10(33), e2305096-e2305096 (2023-10-17)
Despite advances in precision oncology, cancer remains a global public health issue. In this report, proof-of-principle evidence is presented that a cell-penetrable peptide (ACP52C) dissociates transcription factor CP2c complexes and induces apoptosis in most CP2c oncogene-addicted cancer cells through transcription

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service