Skip to Content
Merck
All Photos(1)

Key Documents

SML0046

Sigma-Aldrich

NSC 668036 hydrate

≥98% (HPLC)

Synonym(s):

N-[(1,1-Dimethylethoxy)carbonyl]-L-alanyl-(2S)-2-hydroxy-3-methylbutanoyl-L-Alanine-(1S)-1-carboxy-2-methylpropyl ester hydrate, N-[(1,1-Dimethylethoxy)carbonyl]-L-alanyl-(2S)-2-hydroxy-3-methylbutanoyl-L-alanyl-(2S)-2-hydroxy-3-methylbutanoic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H36N2O9 · xH2O
CAS Number:
Molecular Weight:
460.52 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

H2O: ≥5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O.CC(C)[C@H](OC(=O)[C@H](C)NC(=O)[C@@H](OC(=O)[C@H](C)NC(=O)OC(C)(C)C)C(C)C)C(O)=O

InChI

1S/C21H36N2O9.H2O/c1-10(2)14(30-19(28)13(6)23-20(29)32-21(7,8)9)16(24)22-12(5)18(27)31-15(11(3)4)17(25)26;/h10-15H,1-9H3,(H,22,24)(H,23,29)(H,25,26);1H2/t12-,13-,14-,15-;/m0./s1

InChI key

CARNRIDSPYICIM-WFGXUCIJSA-N

Application

NSC 668036 hydrate may be used in Wnt-mediated cell signaling studies.

Biochem/physiol Actions

Inhibitor of dishevelled PDZ domain.
NSC 668036 binds to the PDZ domain of the Wnt-pathway signaling molecule Disheveled (Dvl), blocking the binding and activation of Dvl by Frizzled. In Xenopus embryos, NSC 668036 inhibited Wnt3A-induced expression of the target gene Siamois and formation of secondary axes.

Other Notes

hygroscopic

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service