Skip to Content
Merck
All Photos(2)

Key Documents

SMB00804

Sigma-Aldrich

D-Gulonic acid lithium salt

Synonym(s):

(2R,3R,4S,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid lithium salt, D-(8CI) Gulonic acid lithium salt, Lithium D-gulonate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€91.30

€91.30


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€91.30

About This Item

Empirical Formula (Hill Notation):
C6H11O7Li
CAS Number:
Molecular Weight:
202.09
UNSPSC Code:
12352205

€91.30


Estimated to ship onMay 14, 2025


Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

storage temp.

2-8°C

SMILES string

O[C@H]([C@@H](O)[C@@H](O)C(=O)O)[C@H](O)CO

InChI

1S/C6H12O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h2-5,7-11H,1H2,(H,12,13)/t2-,3+,4-,5-/m1/s1

InChI key

RGHNJXZEOKUKBD-KKQCNMDGSA-N

General description

D-Gulonic acid lithium salt, also known as Lithium D-gulonate, is a lithium salt of D-Gulonic acid commonly used as a vitamin precursor. D-gulonic acid falls within the category of organic compounds referred to as medium-chain hydroxy acids/sugar acids and derivatives, distinguished by a side chain composed of 6 to 12 carbon atoms. D-gulonic acid inhibits polymerase chain reaction (PCR) and DNA synthesis by binding to the enzyme activities at their active sites, making it a valuable asset in biochemical and metabolomics research.

Application

D-gulonic acid lithium salt can be utilized to investigate microbial metabolism, explore enzyme functional diversity in metabolic processes, characterize metabolic pathways, and understand carbon utilization, providing potential benefits in biotechnology, drug discovery, cell culture, and biomedical research.

Features and Benefits

Versatile and adaptable for a variety of research and metabolomics applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service