Skip to Content
Merck
All Photos(1)

Key Documents

SMB00205

Sigma-Aldrich

Carpachromene

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16O5
CAS Number:
Molecular Weight:
336.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(C)Oc2cc3OC(=CC(=O)c3c(O)c2C=C1)c4ccc(O)cc4

InChI

1S/C20H16O5/c1-20(2)8-7-13-16(25-20)10-17-18(19(13)23)14(22)9-15(24-17)11-3-5-12(21)6-4-11/h3-10,21,23H,1-2H3

InChI key

YXOATFKTEDZPFL-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parham Taslimi
Journal of biochemical and molecular toxicology, 34(11), e22566-e22566 (2020-07-03)
Tyrosinase enzyme is a functional oxidase that is extensively divided in nature. It is the main enzyme in melanin synthesis and is also involved in designating the color of mammalian hair and skin. Additionally, it is accountable for the unfavorable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service