Skip to Content
Merck
All Photos(1)

Key Documents

SAB5202186

Sigma-Aldrich

Monoclonal Anti-Kir6.1-Atto 488 antibody produced in mouse

clone S366-60

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€278.00

€278.00


Estimated to ship onApril 29, 2025



Select a Size

Change View
100 μG
€278.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.45

€278.00


Estimated to ship onApril 29, 2025


biological source

mouse

conjugate

Atto 488 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S366-60, monoclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

human, rat

concentration

1.0 mg/mL

technique(s)

immunofluorescence: 1:100
western blot: 1:1000

isotype

IgG2a

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1

Physical form

PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service