Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700267

Sigma-Aldrich

Monoclonal Anti-CD64 antibody produced in mouse

clone 10.1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FCGR1, Anti-FcγRI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10.1, monoclonal

form

buffered aqueous solution

species reactivity

human, nonhuman primates

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FCGR1(2209)

General description

The mouse monoclonal antibody 10.1 recognizes alpha subunit of CD64/FcgammaRI, a 72 kDa single chain type I glycoprotein, that is expressed on monocytes/macrophages, dendritic cells, and activated granulocytes.

Immunogen

Rheumatoid synovial fluid cells and fibronectin purified human monocytes

Application

Cluster of differentiation 64 (CD64) has been implicated in various immunological processes such as phagocytosis, the clearance of immune complexes and the secretion of immunological mediators. In addition, it also facilitates antibody-dependent cellular cytotoxicity, the modulation of B-cell differentiation and transcytosis of IgG across the epithelial layer. Neutrophil CD64 serves as a potent diagnostic marker for bacterial infection and neonatal sepsis.

Biochem/physiol Actions

The cluster of differentiation 64 (CD64), also known as Fc fragment of IgG receptor Ia (FCGR1A), gene is mapped to human chromosome lq21.2--q21.3. The gene codes for a 72 kDa glycoprotein that is constitutively expressed on human monocytes and macrophages.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neutrophil CD64 as a diagnostic marker for neonatal sepsis: Meta-analysis.
Dai J, et al.
Advances in Clinical and Experimental Medicine : Official Organ Wroclaw Medical University, 327-332 (2017)
Yunqin Lee et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(14), 3571-3581 (2016-02-13)
Human tumors consist of heterogeneous populations of cells with distinct marker expression and functional properties. In squamous cell carcinoma of the head and neck (SCCHN), CD44 is a well-characterized marker of a resilient subpopulation of cells associated with increased tumorigenesis
Neutrophil CD64 expression as a longitudinal biomarker for severe disease and acute infection in critically ill patients.
Jong E, et al.
International Journal of Laboratory Hematology, 38(5), 576-584 (2016)
Human high-affinity Fc?RI (CD64) gene mapped to chromosome 1q21.2-q21.3 by fluorescence in situ hybridization
Takai S, et al.
Human Genetics, 93(1), 13-15 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service