Skip to Content
Merck
All Photos(2)

Key Documents

SAB4301041

Sigma-Aldrich

Anti-CD163 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

M130, MM130

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

2.9 mg/mL

technique(s)

immunohistochemistry: 1:25- 1:100

isotype

IgG

accession no.

NP_004235.4

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD163(9332)

General description

Cluster of differentiation 163 (CD163) is a hemoglobin scavenger receptor, encoded by a gene mapped to human chromosome 12p13.31. It is expressed on the surface of activated macrophages. The encoded protein is a member of scavenger receptor cysteine-rich family (SRCR) and is specifically expressed on cells of the monocyte/macrophage lineage.

Specificity

The antibody detects endogenous levels of total CD163 protein.

Immunogen

Synthetic peptide corresponding to residues near the C terminal of human CD163 molecule

Biochem/physiol Actions

Cluster of differentiation 163 (CD163) modulates anti-inflammatory functions and has a role in hemoglobin-haptoglobin clearance. Various cytokines influence the expression of CD163. It may be associated with hepatocellular carcinoma. Soluble CD163 level in serum acts as a potent indicator of the progression of cirrhosis and portal hypertension.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical significance of soluble CD163 in polymyositis-related or dermatomyositis-related interstitial lung disease.
Enomoto Y, et al.
Arthritis Research & Therapy, 19(1), 9-9 (2017)
Role of Serum Soluble CD163 in the Diagnosis, Risk of Bleeding, and Prognosis of Gastro-Esophageal Varices in Cirrhotic Patients.
Fouad R, et al.
Journal of Interferon & Cytokine Research, 37(3), 112-118 (2017)
SNP selection for genes of iron metabolism in a study of genetic modifiers of hemochromatosis.
Constantine C C, et al.
BMC Medical Genetics, 9(1), 18-18 (2008)
Macrophage activation marker soluble CD163 may predict disease progression in hepatocellular carcinoma.
Kazankov K, et al.
Scandinavian Journal of Clinical and Laboratory Investigation, 76(1), 64-73 (2016)
C Buechler et al.
Journal of leukocyte biology, 67(1), 97-103 (2000-01-27)
CD163, also referred to as M130, a member of the scavenger receptor cysteine-rich family (SRCR) is exclusively expressed on cells of the monocyte lineage. In freshly isolated monocytes the CD14bright CD16+ monocyte subset revealed the highest expression of CD163 among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service