Skip to Content
Merck
All Photos(2)

Documents

SAB4300571

Sigma-Aldrich

Anti-MAP1LC3A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-LC3 antibody produced in rabbit, Anti-LC3A antibody produced in rabbit, Anti-MAP1ALC3 antibody produced in rabbit, Anti-MAP1BLC3 antibody produced in rabbit, Anti-microtubule-associated protein 1 light chain 3 alpha antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~16 kDa
~18 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-P-F-K-Q)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Peptide sequence around aa. 5-10 (R-P-F-K-Q), according to the protein MAP1LC3A.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Probably involved in formation of autophagosomal vacuoles (autophagosomes).

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing Han et al.
Experimental and therapeutic medicine, 22(4), 1054-1054 (2021-08-27)
Human respiratory syncytial virus (RSV) is a major health challenge due to the lack of a safe and effective vaccine and antiviral drugs. RSV non-structural protein 1 (NS1) is the main inhibitor of antiviral signaling pathways in RSV infection; however
Antonio Piras et al.
Cell death & disease, 8(12), 3223-3223 (2017-12-21)
Spinal muscular atrophy (SMA) is a recessive autosomal neuromuscular disease, due to homozygous mutations or deletions in the telomeric survival motoneuron gene 1 (SMN1). SMA is characterized by motor impairment, muscle atrophy, and premature death following motor neuron (MN) degeneration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service