Skip to Content
Merck
All Photos(3)

Key Documents

SAB2702158

Sigma-Aldrich

Monoclonal Anti-RPA3 antibody produced in mouse

Synonym(s):

REPA3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€444.00

€444.00


Estimated to ship onMay 26, 2025



Select a Size

Change View
100 μL
€444.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€444.00


Estimated to ship onMay 26, 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11.1
monoclonal

form

liquid

mol wt

14 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:1000
immunoprecipitation (IP): suitable
western blot: 0.1-2 μg/mL

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPA3(6119)

Immunogen

Full-length human RPA-14 expressed in E. coli..

Application

For IP: Use at a concentration of 0.1 - 2 μg/mL. For WB: Use at a concentration of 0.1 - 2 μg/mL. For IHC-P: 1:100-1:1000. Predicted molecular weight: 14 kDa. Optimal dilutions/concentrations should be determined by the researcher.

Biochem/physiol Actions

Replication protein A (RPA) is a single-stranded DNA binding protein. Human RPA is a heterotrimeric protein containing subunits of 70, 32 and 14 kDa. This protein complex is highly conserved in eukaryotes and is essential in DNA replication, homologous recombination and nucleotide excision repair. RPA has been shown to be a target for DNA-dependent protein kinase (DNA-PK) ataxia telangiectasia-mutated gene (ATM) and a cyclin dependent protein kinase.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service