Skip to Content
Merck
All Photos(2)

Key Documents

SAB2500458

Sigma-Aldrich

Anti-GCNT3 (AA 410-422) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-C2/4GnT, Anti-C2GnT-M, Anti-C2GnT2, Anti-N-Acetylglucosaminyltransferase 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€419.00

€419.00


Check Cart for Availability


Select a Size

Change View
100 μG
€419.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€419.00


Check Cart for Availability

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse, canine

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCNT3(9245)

General description

The GCNT3 (glucosaminyl (N-acetyl) transferase 3, mucin type) gene codes for an enzyme synthesizing the core of mucin protein and is mapped to human chromosome 15q22.2. The enzyme is predominantly expressed in stomach and colon.

Immunogen

Peptide with sequence C-NKFDPKVDDNALQ from the internal region (near C terminus) of the protein sequence according to NP_004742.1.

Biochem/physiol Actions

Gcnt3 (glucosaminyl (N-acetyl) transferase 3, mucin type) is involved in the biosynthesis of core 2, core 4 and blood group I antigen in secreted mucins. Overexpression of GCNT3 gene is observed in human pancreatic cancer and contributes to elevated mucin formation, aggressive tumorigenesis and decreased patient survival. The GCNT3 gene is known to promote cell proliferation and spheroid formation in tumors. Deletion of GCNT3 gene results in loss of core 2 O-glycan in the stomach and colon.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Core 2 Mucin-Type O-Glycan Is Related to EPEC and EHEC O157:H7 Adherence to Human Colon Carcinoma HT-29 Epithelial Cells.
Jun Y, et al.
Digestive Diseases and Sciences, 60(7), 1977?1990-1977?1990 (2015)
Allelic Skewing of DNA Methylation Is Widespread across the Genome.
Leonard C S, et al.
American Journal of Human Genetics, 86(2), 196-212 (2010)
Keratin 1 Plays a Critical Role in Golgi Localization of Core 2 N-Acetylglucosaminyltransferase M via Interaction with Its Cytoplasmic Tail.
Armen P, et al.
The Journal of Biological Chemistry, 290(10), 6256-6269 (2015)
Molecular Pathways: Mucins and Drug Delivery in Cancer.
Chinthalapally V R, et al.
Clinical Cancer Research (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service