Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500189

Sigma-Aldrich

Anti-CAMK1D antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CKLiK, Anti-CaM-K1, Anti-CaMKID, Anti-Calcium/calmodulin-dependent protein kinase, Anti-RP11-462F15.1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€419.00

€419.00


Estimated to ship onApril 29, 2025



Select a Size

Change View
100 μG
€419.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€419.00


Estimated to ship onApril 29, 2025


biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK1D(57118)

General description

The calcium/calmodulin-dependent protein kinase 1D (CAMK1D) gene codes for a member of the Ca2+/calmodulin-dependent protein kinase 1 subfamily of serine/threonine kinases. It is located on human chromosome 10p13.

Immunogen

Peptide with sequence ASQKDCAYVAKPES from the C Terminus of the protein sequence according to NP_065130.1.

Biochem/physiol Actions

Calcium/calmodulin-dependent protein kinase 1D (CAMK1D) modulates the role of granulocytes through chemokine signaling transduction pathway. It is associated with type 2 diabetes (T2D).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation into the promoter DNA methylation of three genes (CAMK1D, CRY2 and CALM2) in the peripheral blood of patients with type 2 diabetes
J Cheng, et al.
Experimental and Therapeutic Medicine, 8(2), 579-584 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service