Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102610

Sigma-Aldrich

Anti-TUT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ21850, Anti-FLJ22267, Anti-FLJ22347, Anti-MGC131987, Anti-Terminal uridylyl transferase 1, U6 snRNA-specific

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Estimated to ship onMay 15, 2025



Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Estimated to ship onMay 15, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

94 kDa

species reactivity

guinea pig, bovine, human, horse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUT1(64852)

General description

The previously assigned protein identifier A8K995 has been merged into Q9H6E5. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the N terminal region of human TUT1

Biochem/physiol Actions

TUT1 is a nucleotidyl transferase that functions as both a terminal uridylyltransferase and a nuclear poly(A) polymerase. TUT1 specifically adds and removes nucleotides from the 3′ end of small nuclear RNAs and select mRNAs and may function in controlling gene expression and cell proliferation.TUT1 specifically catalyzes uridylylation of U6 snRNA (RNU6; MIM 180692) and is essential for cell proliferation (Trippe et al., 2006 [PubMed 16790842]).[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: CDLDLFLDLGDLEEPQPVPKAPESPSLDSALASPLDPQALACTPASPPDS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily C Knouf et al.
PloS one, 8(7), e69630-e69630 (2013-07-23)
Post-transcriptional modifications of miRNAs with 3' non-templated nucleotide additions (NTA) are a common phenomenon, and for a handful of miRNAs the additions have been demonstrated to modulate miRNA stability. However, it is unknown for the vast majority of miRNAs whether

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service