Skip to Content
Merck
All Photos(1)

Key Documents

SAB2100062

Sigma-Aldrich

Anti-ADNP antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ADNP1, Anti-Activity-dependent neuroprotector homeobox, Anti-KIAA0784

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Check Cart for Availability


Select a Size

Change View
100 μL
€498.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

124 kDa

species reactivity

dog, human, rabbit, horse, bovine, mouse, guinea pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADNP(23394)

Immunogen

Synthetic peptide directed towards the N terminal region of human ADNP

Biochem/physiol Actions

Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. ADNP is a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. It contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. It is also upregulated in normal proliferative tissues. Finally, ADNP may increase the viability of certain cell types through modulation of p53 activity.Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.

Sequence

Synthetic peptide located within the following region: EDFKQFEPNDFYLKNTTWEDVGLWDPSLTKNQDYRTKPFCCSACPFSSKF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service