Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402522

Sigma-Aldrich

Monoclonal Anti-PITRM1 antibody produced in mouse

clone 1H3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIAA1104, MGC138192, MGC141929, MP1, PreP, hMP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€490.00

€490.00


Estimated to ship onApril 27, 2025



Select a Size

Change View
100 μG
€490.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€490.00


Estimated to ship onApril 27, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~84.48 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PITRM1(10531)

Related Categories

Immunogen

PITRM1 (AAH01150, 1 a.a. ~ 534 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MRPDDKYHEKQAQVEATKLKQKVEALSPGDRQQIYEKGLELRSQQSKPQDASCLPALKVSDIEPTIPVTELDVVLTAGDIPVQYCAQPTNGMVYFRAFSSLNTLPEELRPYVPLFCSVLTKLGCGLLDYREQAQQIELKTGGMSASPHVLPDDSHMDTYEQGVLFSSLCLDRNLPDMMQLWSEIFNNPCFEEEEHFKVLVKMTAQELANGIPDSGHLYASIRAGRTLTPAGDLQETFSGMDQVRLMKRIAEMTDIKPILRKLPRIKKHLLNGDNMRCSVNATPQQMPQTEKAVEDFLRSIGRSKKERRPVRPHTVEKPVPSSSGGDAHVPHGSQVIRKLVMEPTFKPWQMKTHFLMPFPVNYVGECIRTVPYTDPDHASLKILARLMTAKFLHTEIREKGGAYGGGAKLSHNGIFTLYSYRDPNTIETLQSFGKAVDWAKSGKFTQQDIDEAKLSVFSTVDAPVAPSDKGMDHFLYGLSDEMKQAHREQLFAVSHDKLLAVSDRYLGTGKSTHGLAILGPENPKIAKDPSWIIR

Biochem/physiol Actions

PITRM1 (Pitrilysin metallopeptidase 1) acts as a mitochondrial amyloid-β peptide (Aβ)-degrading enzyme localized in the mitochondrial matrix. Its activity is controlled by Zn(+2)-dependent metalloprotease inhibitor. The temporal lobe of the human brain has a high tendency of Aβ accumulation and reactive oxygen species (ROS) production. In Alzheimer′s disease, the low proteolytic activity of PITRM1 in the brain mitochondria has been reported. Studies have been suggested that high ROS production results in decreased PITRM1 activity followed by Aβ accumulation. As a result, it leads to the mitochondrial toxicity and neuronal death. Thus, it proves the impact of PITRM1 in the Alzheimer′s disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nyosha Alikhani et al.
Journal of Alzheimer's disease : JAD, 27(1), 75-87 (2011-07-14)
Accumulation of amyloid-β peptide (Aβ), the neurotoxic peptide implicated in the pathogenesis of Alzheimer's disease (AD), has been shown in brain mitochondria of AD patients and of AD transgenic mouse models. The presence of Aβ in mitochondria leads to free
N Mzhavia et al.
DNA and cell biology, 18(5), 369-380 (1999-06-09)
A novel cDNA, designated human metalloendoprotease 1 (hMP1), was identified on the basis of homology to known metalloendoproteases of the pitrilysin family. The full-length MP1 codes for a protein with an open reading frame of 1038 amino acids. The N-terminal
Catarina Moreira Pinho et al.
Neuroscience letters, 469(2), 204-208 (2009-12-08)
Several studies suggest mitochondrial dysfunction as a possible mechanism underlying the development of Alzheimer disease (AD). There is data showing that amyloid-beta (A beta) peptide is present in AD brain mitochondria. The human presequence protease (hPreP) was recently shown to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service