Skip to Content
Merck
All Photos(1)

Key Documents

S8201

Sigma-Aldrich

SR-3576

≥98% (HPLC)

Synonym(s):

3-(5-(3-m-Tolylureido)-1H-indazol-1-yl)-N-(3,4,5-trimethoxyphenyl)benzamide, 3-[4-[[[(3-Methylphenyl)amino]carbonyl]amino]-1H-pyrazol-1-yl]-N-(3,4,5-trimethoxyphenyl)-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27N5O5
CAS Number:
Molecular Weight:
501.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

light brown to brown

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc(NC(=O)c2cccc(c2)-n3cc(NC(=O)Nc4cccc(C)c4)cn3)cc(OC)c1OC

InChI

1S/C27H27N5O5/c1-17-7-5-9-19(11-17)30-27(34)31-21-15-28-32(16-21)22-10-6-8-18(12-22)26(33)29-20-13-23(35-2)25(37-4)24(14-20)36-3/h5-16H,1-4H3,(H,29,33)(H2,30,31,34)

InChI key

MTFAYLZZDJGFGV-UHFFFAOYSA-N

Application

SR-3576 may be used in JNK3-mediated cell signaling studies.

Biochem/physiol Actions

SR-3576 is a cell-permeable, potent and selective JNK3 (c-Jun N-terminal kinase 3) inhibitor.
SR-3576 is a small molecule inhibitor belonging to aminopyrazole class that selectively inhibits JNK3 with >2800 fold selectively over p38.1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ted Kamenecka et al.
The Journal of biological chemistry, 284(19), 12853-12861 (2009-03-06)
c-Jun N-terminal kinase 3alpha1 (JNK3alpha1) is a mitogen-activated protein kinase family member expressed primarily in the brain that phosphorylates protein transcription factors, including c-Jun and activating transcription factor-2 (ATF-2) upon activation by a variety of stress-based stimuli. In this study
Sara Barreto et al.
Acta biomaterialia, 53, 59-69 (2017-02-22)
In order to identify the mechanisms by which skeletal maturity alters the mechanosensitivity of mesenchymal stromal cells (MSCs) and, the implications for osteogenesis and angiogenesis during bone formation, we compared the response of MSCs derived from children and skeletally-mature healthy
Mei-Chou Lai et al.
Nutrients, 15(3) (2023-02-12)
Degenerative diseases of the brain include Parkinson's disease (PD), which is associated with moveable signs and is still incurable. Hispidin belongs to polyphenol and originates primarily from the medicinal fungi Inonotus and Phellinus, with distinct biological effects. In the study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service