Key Documents
RF454811P-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-48-PCR, to hold, 6 x 8-tube or 5-tube PCR strips, to hold, 48 x 0.2 mL PCR tubes
About This Item
UNSPSC Code:
41103909
Pricing and availability is not currently available.
feature
to hold 48 x 0.2 mL PCR tubes
to hold 6 x 8-tube or 5-tube PCR strips
fixed angle strip rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022638581 (NA)
Eppendorf® 5804735005 (EU)
parameter
15,350 × g max. RCF
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)