R2027
Ral B human
recombinant, expressed in E. coli, buffered aqueous glycerol solution
Select a Size
€803.00
Estimated to ship onMay 22, 2025
Select a Size
About This Item
€803.00
Estimated to ship onMay 22, 2025
recombinant
expressed in E. coli
Quality Level
form
buffered aqueous glycerol solution
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... RALB(5899)
Application
Biochem/physiol Actions
Other Notes
Physical form
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service