Skip to Content
Merck
All Photos(1)

Key Documents

R1638

Cryogenic storage rack, stainless steel

Vertical, 4-shelf for 3 × 3 × 2 in. boxes

Synonym(s):

cryogenic vial rack, cryostorage rack, cryovial rack, dewar rack, liquid nitrogen rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€267.00

€267.00


Check Cart for Availability


Select a Size

Change View
1 EA
€267.00

About This Item

UNSPSC Code:
41122806
NACRES:
NB.22

€267.00


Check Cart for Availability

material

stainless steel

feature

Holds 12 x boxes, 3 × 3 × 2 in.

packaging

pack of 1 ea

manufacturer/tradename

Nalgene® DS5035-0004

technique(s)

cryopreservation: suitable

Overall H

8 3/4 in.

Looking for similar products? Visit Product Comparison Guide

General description

Racks hold CryoBoxes securely but separately, allowing fast retrieval and immediate return to freezer. The vertical racks are best for use in chest freezers, horizontal racks for upright freezers.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service