Skip to Content
Merck
All Photos(1)

Key Documents

PZ0269

Sigma-Aldrich

PF-03049423

≥98% (HPLC)

Synonym(s):

3-[4-(2-Hydroxyethyl)-1-piperazinyl]-7-(6-methoxy-3-pyridinyl)-1-(2-propoxyethyl)-pyrido[3,4-b]pyrazin-2(1H)-one hydrochloride, 3-[4-(2-Hydroxyethyl)piperazin-1-yl]-7-(6-methoxypyridin-3-yl)-1-(2-propoxyethyl)pyrido[3,4-b]pyrazin-2(1H)-one hydrochloride, PF-5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N6O4 · xHCl
CAS Number:
Molecular Weight:
468.55 (free base basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CCCOCCN1C2=C(C=NC(C3=CN=C(OC)C=C3)=C2)N=C(N4CCN(CCO)CC4)C1=O.C

InChI

1S/C24H32N6O4.CH4/c1-3-13-34-14-11-30-21-15-19(18-4-5-22(33-2)26-16-18)25-17-20(21)27-23(24(30)32)29-8-6-28(7-9-29)10-12-31;/h4-5,15-17,31H,3,6-14H2,1-2H3;1H4

InChI key

BXMDZHBYGYPATN-UHFFFAOYSA-N

Biochem/physiol Actions

PF-03049423 is an orally active, brain penetrant phosphodiesterase 5 (PDE5) inihbitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service