Skip to Content
Merck
All Photos(2)

Key Documents

OGS1634

Sigma-Aldrich

PSF-TEF1-NH2-HA-TEV - N-TERMINAL HA TAG YEAST PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

tag

Hemagglutinin A (HA) tagged

form

buffered aqueous solution

mol wt

size 6763 bp

bacteria selection

kanamycin

Origin of replication

2Micron
pUC (500 copies)

Peptide cleavage

TEV

Peptide tag location

N-terminal

Promoter

Promoter name: TEF1
Promoter activity: constitutive
Promoter type: yeast

reporter gene

none

shipped in

ambient

storage temp.

−20°C

yeast selection

uracil

General description

This plasmid is designed to express tagged proteins in yeast cells (Saccharomyces cerevisiae). The plasmid contains an auxotrophic Uracil selection expression cassette (URA3) that allows for the positive selection of yeast that are deficient in the URA3 gene (YEL021W). This is typically achieved by growing the yeast in minimal media that is reconstituted with the essential amino acids and nucleotides but excluding Uracil.

About the Peptide Tag:This plasmid contains an n-terminal Influenza Hemagglutinin (HA) epitope tag that can be fused to a gene of interest to allow protein detection and/or purification. The sequence of the tag is: YPYDVPDYA.

About the Cleavage Tag:This plasmid also encodes a protease cleavage site that is designed to be positioned between your gene of interest and the tag to allow the removal of the tag following protein purification or isolation. This plasmid contains a TEV cleavage tag. The protein sequence of the cleavage tag is: ENLYFQG. Cleavage occurs between the Glu and Gly residues. TEV is often reported to have better specificity for its recognition site compared to EKT Thrombin or Faxtor Xa.

Promoter Expression Level: This plasmid contains the Yeast Elongation Factor Alpha promoter (TEF1). This is the strongest of the yeast promoters that we sell. It is a constitutive promoter and requires no induction. If you are interested in weaker promoters levels than we also stock plasmids that contain the following promoters in order of decreasing strength TPI (strong), ADHI (medium), STE5 (weak). We also stock Galactose inducible promoter plasmids if inducible expression is required. Please contact us for further information.

Sequence

To view sequence information for this product, please visit the product page

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer
Alexander C Cerny et al.
PLoS genetics, 11(10), e1005578-e1005578 (2015-10-29)
Recycling of signaling proteins is a common phenomenon in diverse signaling pathways. In photoreceptors of Drosophila, light absorption by rhodopsin triggers a phospholipase Cβ-mediated opening of the ion channels transient receptor potential (TRP) and TRP-like (TRPL) and generates the visual
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown.
Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service