MOU964
Mouse Oligonucleotide Libraries
(96 well) plate 4
Select a Size
€7,550.00
Estimated to ship onApril 18, 2025
Select a Size
About This Item
€7,550.00
Estimated to ship onApril 18, 2025
Recommended Products
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
• Libraries are arranged by gene function and are available as an entire library or subsets of the library in 4x96- and 384-well plate format
• Quality tested using PAGE, Mass Spec., and capillary electrophoresis to verify purity, synthesis, length and orientation in plates
• Includes access to LabOnWeb.com where you will find detailed information about the oligos and genes, including accession numbers, sequence information, oligo properties, gene annotation, and more
Packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service