Skip to Content
Merck
All Photos(1)

Documents

MAK484

Sigma-Aldrich

Phenylalanine Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

Assay Kit for Phenylalanine Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

detection method

fluorometric

relevant disease(s)

metabolic disorders; neonatal diseases; pediatric diseases; neurological disorders

shipped in

wet ice

storage temp.

−20°C

Related Categories

General description

Phenylalanine is an essential amino acid and an important precursor for several key signal molecules such as dopamine, norepinephrine, epinephrine, and the skin pigment melanin. It is found naturally in the breast milk of mammals and used as nutritional supplements in food and drink products. The genetic disorder phenylketonuria is the inability to metabolize phenylalanine. Individuals with this disorder are known as "phenylketonurics". Individuals who cannot metabolize phenylalanine must monitor their intake of protein to control the buildup of phenylalanine.

Features and Benefits

  • The linear detection range of the kit is 2-300 µM L-phenylalanine
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for L-phenylalanine activity determination in serum, urine, and other biological samples.

Principle

The Phenylalanine Assay Kit provides a convenient fluorometric means to measure L-phenylalanine in biological samples. In the assay, L-phenylalanine is oxidized by phenylalanine dehydrogenase, producing NADH, which reduces a fluorescent dye to a highly fluorescent product. The resulting fluorescence intensity at λEx = 530 nm/λEm = 585 nm is proportional to the L-phenylalanine concentration in the sample.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service