Skip to Content
Merck
All Photos(2)

Key Documents

M9519

Sigma-Aldrich

4-Methylumbelliferyl α-L-arabinofuranoside

glycosidase substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O7
CAS Number:
Molecular Weight:
308.28
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (TLC)

form

powder

impurities

≤10% methanol

solubility

pyridine: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(OC3OC(CO)C(O)C3O)ccc12

InChI

1S/C15H16O7/c1-7-4-12(17)21-10-5-8(2-3-9(7)10)20-15-14(19)13(18)11(6-16)22-15/h2-5,11,13-16,18-19H,6H2,1H3

InChI key

FAGLTVBWEMHJRP-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 1

Target Organs

Eyes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna M Alessi et al.
Environmental microbiology, 22(6), 2150-2164 (2020-03-07)
A clone encoding carboxymethyl cellulase activity was isolated during functional screening of a human gut metagenomic library using Lactococcus lactis MG1363 as heterologous host. The insert carried a glycoside hydrolase family 9 (GH9) catalytic domain with sequence similarity to a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service