Skip to Content
Merck
All Photos(1)

Key Documents

M3376

Supelco

2-(4-Methoxyphenyl)ethylamine

liquid

Synonym(s):

4-Methoxyphenethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
151.21
Beilstein:
508967
EC Number:
MDL number:
UNSPSC Code:
41116107

form

liquid

drug control

Home Office Schedule 1

color

light yellow

refractive index

n20/D 1.538 (lit.)

bp

138-140 °C/20 mmHg (lit.)
254-256 °C

density

1.031 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(CCN)cc1

InChI

1S/C9H13NO/c1-11-9-4-2-8(3-5-9)6-7-10/h2-5H,6-7,10H2,1H3

InChI key

LTPVSOCPYWDIFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite of tyramine

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service