Skip to Content
Merck
All Photos(1)

Key Documents

M3140

Sigma-Aldrich

Methyl 6-deoxy-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O5
CAS Number:
Molecular Weight:
178.18
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

COC1OC(C)C(O)C(O)C1O

InChI

1S/C7H14O5/c1-3-4(8)5(9)6(10)7(11-2)12-3/h3-10H,1-2H3

InChI key

OHWCAVRRXKJCRB-UHFFFAOYSA-N

Application

Methyl 6-deoxy-β-D-glucopyranoside has been used in a study to advance the understanding of the differences in reactivity of single hydroxyl groups on the pyranoside skeleton.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Partial tosylation of methyl 6-deoxy-β-D-glucopyranoside
Stanek, J., et al.
Collection of Czechoslovak Chemical Communications, 40, 3370-3385 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service