Skip to Content
Merck
All Photos(1)

Key Documents

M1768

Sigma-Aldrich

1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole

vial of 1.78 g (6 mmol)

Synonym(s):

(2,4,6-Trimethylphenyl)-(3-nitro-1,2,4-triazol-1-yl) sulfone, MSNT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O4S
CAS Number:
Molecular Weight:
296.30
Beilstein:
4328921
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

packaging

vial of 1.78 g (6 mmol)

mp

134-139 °C (lit.)

storage temp.

−20°C

SMILES string

Cc1cc(C)c(c(C)c1)S(=O)(=O)n2cnc(n2)[N+]([O-])=O

InChI

1S/C11H12N4O4S/c1-7-4-8(2)10(9(3)5-7)20(18,19)14-6-12-11(13-14)15(16)17/h4-6H,1-3H3

InChI key

SFYDWLYPIXHPML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Condensing reagent for oligonucleotide synthesis.

Packaging

Packaged under argon and sealed with PTFE-faced septum.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service