Skip to Content
Merck
All Photos(2)

Key Documents

L9754

Sigma-Aldrich

Linoleoyl coenzyme A lithium salt

≥85%

Synonym(s):

(cis,cis)-9,12-Octadecadienoyl Coenzyme A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H66N7O17P3S
CAS Number:
Molecular Weight:
1029.96
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥85%

storage temp.

−20°C

SMILES string

[Li].CCCCC\C=C\C\C=C\CCCCCCCC(=O)SCCNC(=O)CCNC(=O)C(O)C(C)(C)COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1OP(O)(O)=O)n2cnc3c(N)ncnc23

InChI

1S/C39H66N7O17P3S.Li.H/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-30(48)67-23-22-41-29(47)20-21-42-37(51)34(50)39(2,3)25-60-66(57,58)63-65(55,56)59-24-28-33(62-64(52,53)54)32(49)38(61-28)46-27-45-31-35(40)43-26-44-36(31)46;;/h8-9,11-12,26-28,32-34,38,49-50H,4-7,10,13-25H2,1-3H3,(H,41,47)(H,42,51)(H,55,56)(H,57,58)(H2,40,43,44)(H2,52,53,54);;/b9-8+,12-11+;;

InChI key

CQYVZXLGBQGVMO-VTXMMSKZSA-N

Biochem/physiol Actions

Coenzyme A functions as an acyl group carrier, acetyl-CoA. Linoleoyl coenzyme A is a substrate used to study the specificity and kinetics of mitochondrial linoleoyl-coenzyme monolysocardiolipin acyltransferase-1 (MLCL AT-1) and acyl-CoA: lysophosphatidylcholine acyltransferase (LPCAT).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mauricio A Retamal et al.
Pflugers Archiv : European journal of physiology, 461(6), 635-643 (2011-03-02)
Connexins form hemichannels at undocked plasma membranes and gap-junction channels (GJCs) at intercellular contacting zones. Under physiological conditions, hemichannels have low open probabilities, but their activation under pathological conditions, such as ischemia, induces and/or accelerates cell death. Connexin 46 (Cx46)
Michael Kazachkov et al.
Lipids, 43(10), 895-902 (2008-09-11)
An important enzyme involved in phospholipid turnover is the acyl-CoA: lysophosphatidylcholine acyltransferase (LPCAT). Here, we report characterization of a newly discovered human LPCAT (LPCAT3), which has distinct substrate preferences strikingly consistent with a role in phosphatidylcholine (PtdCho) remodeling and modulating
William A Taylor et al.
The Journal of biological chemistry, 284(44), 30360-30371 (2009-09-10)
Here we report the identification of a previously uncharacterized human protein as the human monolysocardiolipin acyltransferase-1 (MLCL AT-1). Pig liver mitochondria were treated with n-butyl alcohol followed by Q-Sepharose chromatography, preparative gel electrophoresis, cytidine diphosphate-1,2-diacyl-sn-glycerol-Sepharose chromatography, and finally monolysocardiolipin-adriamycin-agarose affinity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service