Skip to Content
Merck
All Photos(4)

Documents

I17003

Sigma-Aldrich

Interleukin-4 human

IL-4, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested

Synonym(s):

Interleukin-4 human, IL-4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥98% (SDS-PAGE)

potency

≤1 ng/mL ED50

mol wt

14.9 kDa (glycosylated)

technique(s)

cell culture | mammalian: suitable

suitability

endotoxin tested

storage temp.

−20°C

General description

Recombinant human Interleukin-4 (IL-4) is expressed in human 293 cells as a glycoprotein with a calculated moleculaer mass of 14.9 kDa. This protein is manufactured in human cells using an all-human production system, with full chemically defined ingredients and with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports better stability of the protein in culture.

Biochem/physiol Actions

IL-4 is a protein that has been shown to regulate a wide spectrum of functions in B cells, T cells, monocytes/macrophages and other haematopoietic and non-haematopoietic cells. Among T cell clones, IL-4 is produced by Th0 and Th2 cells, but not Th1 cells, and this has now been demonstrated both in mice and in humans. IL-4 blocks the production of proinflammatory cytokines such IL-6, TNFα, and M-CSF and improves the differentiation of immature DCs from CD34+ progenitors when added during the differentiation period (day 6 to day 12).
Interleukin-4 is a lymphokine with profound effects on the growth and differentiation of immunologically competent cells. IL-4 is also known as B cell stimulatory factor-1 (BSF-1), T cell growth factor-2 (TCGF-2) and mast-cell growth factor-2 (MCGF-2). Inhibits VEGF-induced and bFGF-induced angiogenesis. IL-4 is a complex glycoprotein released by a subset of activated T cells. Human and mouse IL-4 share 50% amino acid sequence homology, but their biological actions are species-specific.

Sequence

HKCDITLQEIIKTLNSLTEQKTLCTELTVTDIFAASKNTTEKETFCRAATVLRQFYSHHEKDTRCLGATAQQFHRHKQLIRFLKRLDRNLWGLAGLNSCPVKEANQSTLENFLERLKTIMREKYSKCSS

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Analysis Note

The biological activity of recombinant human IL-4 was tested in culture by measuring its ability to stimulate proliferation of human TF-1 cells (human erythroleukemic indicator cell line).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service