HUTR00441
MISSION® 3′UTR Lenti GoClone™
Powered by SwitchGear Genomics™, 3′UTR, human, SLC6A18
Synonym(s):
HUTR
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
Functional Validation of miRNA Gene Targets
product line
MISSION®
concentration
≥1x105 VP/ml (via p24 assay)
human 3′UTR sequence
CTTGCTCAGCTGCTCACCCGGCGGAGGCGGACGTGGAGGGACAGGGACGCGCGCCCAGACACGGACATGCGCCCGGACACGGACACGCGCCCAGACACGG
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... SLC6A18(348932)
Application
To see more application data, protocols, and vector maps visit www.sigma.com/3utr.
Physical form
200 μL of at least 105 VP/mL (via p24 titering assay) lentiviral particles are provided as frozen stock.
Legal Information
Use of this product is subject to one or more license agreements. For details, please see www.sigmaaldrich.com/missionlicense.
GoClone is a trademark of SwitchGear Genomics
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
SwitchGear Genomics is a trademark of SwitchGear Genomics
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service