Skip to Content
Merck
All Photos(4)

Key Documents

HPA006960

Sigma-Aldrich

Anti-LITAF antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LPS-induced TNF-α factor antibody produced in rabbit, Anti-Lipopolysaccharide-induced tumor necrosis factor-α factor antibody produced in rabbit, Anti-Small integral membrane protein of lysosome/late endosome antibody produced in rabbit, Anti-p53-induced protein 7 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€490.00

€490.00


Check Cart for Availability


Select a Size

Change View
100 μL
€490.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€490.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

APPSYEETVAVNSYYPTPPAPMPGPTTGLVTGPDGKGMNPPSYYTQPAPIPNNNPITVQTVYVQHPITFLDRPIQMCCPSCNKMIVSQLSYNAGALTWLSCGSLCLLGCIAGCCFIPFCVDALQDVDHYCPNC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LITAF(9516)

Immunogen

Lipopolysaccharide-induced tumor necrosis factor-α factor recombinant protein epitope signature tag (PrEST)

Application

Anti-LITAF antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

LITAF (Lipopolysaccharide-induced TNF factor) is a widely expressed integral membrane protein of lysosome/late endosome. It activates monocytes and macrophages, which further accelerates the tumor necrosis factor α (TNF-α) secretion and other inflammatory mediators. It is localized in the intraluminal vesicles of multivesicular bodies (MVBs) and inside exosomes. It is involved in the generation of exosomes through modulating the MVBs formation. Mutation in LITAF causes a rare neurological autosomal-dominant demyelination disorder in CMT1C (Charcot-Marie-Tooth 1C).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71014

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Myokai et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(8), 4518-4523 (1999-04-14)
Lipopolysaccharide (LPS) is a potent stimulator of monocytes and macrophages, causing secretion of tumor necrosis factor alpha (TNF-alpha) and other inflammatory mediators. Given the deleterious effects to the host of TNF-alpha, it has been postulated that TNF-alpha gene expression must
Wenjing Li et al.
Molecular and cellular biology, 35(14), 2464-2478 (2015-05-13)
Endosomal trafficking is a key mechanism to modulate signal propagation and cross talk. Ubiquitin adaptors, along with endosomal sorting complex required for transport (ESCRT) complexes, are also integrated to terminate ligand-receptor activation in late endosomes and multivesicular bodies (MVBs). Within
Hong Zhu et al.
Molecular biology of the cell, 24(11), 1619-1637 (2013-04-12)
Charcot-Marie-Tooth (CMT) disease is an inherited neurological disorder. Mutations in the small integral membrane protein of the lysosome/late endosome (SIMPLE) account for the rare autosomal-dominant demyelination in CMT1C patients. Understanding the molecular basis of CMT1C pathogenesis is impeded, in part

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service