Skip to Content
Merck
All Photos(6)

Key Documents

HPA001616

Sigma-Aldrich

Anti-KNG1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BDK, Anti-BK, Anti-KNG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ALKKYNSQNQSNNQFVLYRITEATKTVGSDTFYSFKYEIKEGDCPVQSGKTWQDCEYKDAAKAATGECTATVGKRSSTKFSVATQTCQITPAEGPVVTAQYDCLGCVHPISTQSPDLEPILRHGIQYFNNN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KNG1(3827)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Kininogen-1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-KNG1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

KNG1 (Kininogen-1) gene encodes a protein that inhibits thiol proteases. Alternative splicing of this protein results in two proteins, high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is involved in coagulation of blood and in the assembly of the kallikrein-kinin system. Apart from being an inhibitor, HMWK also serves as a substrate of platelet calpain. Bradykinin, an active peptide released from HMWK, function in several processes such as smooth muscle contraction, hypotension, natriuresis and diuresis, etc. It also exhibits antibacterial and antifungal activity. KNG-1 acts as a serum biomarker for advanced colorectal adenoma and colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84528

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prestige Antibodies-monospecific antibodies designed for immunohistochemical analysis.
Sara A Gunnerås et al.
BioTechniques, 44(4), 573-576 (2008-05-15)
Jing Wang et al.
PloS one, 8(7), e70519-e70519 (2013-07-31)
Serum markers represent potential tools for the detection of colorectal cancer (CRC). The aim of this study was to obtain proteomic expression profiles and identify serum markers for the early detection of CRC. Proteomic profiles of serum samples collected from
A H Schmaier et al.
The Journal of clinical investigation, 77(5), 1565-1573 (1986-05-01)
Recent studies from our laboratory indicate that a high concentration of platelet-derived calcium-activated cysteine protease (calpain) can cleave high molecular weight kininogen (HMWK). On immunodiffusion and immunoblot, antiserum directed to the heavy chain of HMWK showed immunochemical identity with alpha-cysteine
Karla Jarkovska et al.
Molecular human reproduction, 17(11), 679-692 (2011-06-24)
Ovarian hyperstimulation syndrome (OHSS) is an iatrogenic complication and potentially life-threatening condition resulting from excessive ovarian stimulation during assisted reproductive technologies. Our aim was to identify candidate proteins in follicular fluid (FF) using various proteomic approaches which may help to
Anthony Williams et al.
Transfusion and apheresis science : official journal of the World Apheresis Association : official journal of the European Society for Haemapheresis, 29(3), 255-258 (2003-10-24)
An example of an approach to the developmental philosophy of novel recombinant products is explored by using the exemplar of Hereditary Angioedema (HAE). Plasma kallikrein is believed to be an important mediator of angioedema in patients with genetic deficiency of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service